missing translation for 'onlineSavingsMsg'
Learn More

Cytiva HyClone™ SFM4HEK293 Powdered Media

Catalog No. SH3052201
Change view
Click to view available options
Quantity:
5 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SH3052201 5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SH3052201 Supplier Cytiva Supplier No. SH30522.01

Promotes adenovirus and recombinant protein production in HEK 293 cell culture.

  • Optimized for high cell growth and high recombinant protein and vector production with human embryonic kidney (HEK) 293 cells
  • Protein-free and animal-derived component-free medium available in liquid and powder form
  • Designed for small- and large-scale biomanufacturing processes
  • Manufactured in cGMP (21 CFR 820) compliant and ISO9001 certified facilities
  • Supports direct or sequential adaptation

Specifications

Content And Storage 2 to 8°C, away from light and moisture
Form Powder
Sterility Non-sterile
Without Additives No Glutamine, No Phenol Red, No Poloxamer 188, No Sodium Bicarbonate
Quantity 5 L