missing translation for 'onlineSavingsMsg'
Learn More

Cytiva HyClone™ PF-CHO™ LS and PF-CHO MPS Media

Catalog No. SH3035902
Change view
Click to view available options
Quantity:
1000 mL
5 L
With Additives:
Glutamine, Poloxamer 188, Sodium Bicarbonate
None
Without Additives:
No Glutamine, No Phenol Red, No Poloxamer 188, No HEPES, No Sodium Bicarbonate
No Phenol Red, No HEPES
Product Type:
PF-CHO LS
PF-CHO MPS
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity With Additives Without Additives Product Type
SH3035902 1000 mL Glutamine, Poloxamer 188, Sodium Bicarbonate No Phenol Red, No HEPES PF-CHO LS
SH3033301 5 L None No Glutamine, No Phenol Red, No Poloxamer 188, No HEPES, No Sodium Bicarbonate PF-CHO MPS
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. SH3035902 Supplier Cytiva Supplier No. SH30359.02
NLSU: Get 1 Consumable Product When You Buy 1 Obtenez 1 produit consommable à l’achat d’un produit
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Support growth of multiple CHO cell clones and production of a variety of recombinant proteins. HyClone™ PF CHO™ LS and PF CHO MPS Media is a protein-free media developed through Metabolic Pathway Design.

  • Designed for high cell yield and recombinant protein production
  • Allow for direct or sequential adaptation
  • Support growth in multiple cell culture systems
  • Manufactured from traceable components according to cGMP (21 CFR 820) guidelines
  • Specifications

    Form Liquid
    Product Type PF-CHO LS
    Sterilization Method 0.2 μm Sterile-filtered
    With Additives Glutamine, Poloxamer 188, Sodium Bicarbonate
    Without Additives No Phenol Red, No HEPES
    Quantity 1000 mL
    Certifications/Compliance cGMP
    Content And Storage 2° to 8°C, away from light