missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva HyClone™ PF-CHO™ LS and PF-CHO MPS Media
Support growth of multiple CHO cell clones and production of a variety of recombinant proteins. HyClone™ PF CHO™ LS and PF CHO MPS Media is a protein-free media developed through Metabolic Pathway Design.
Supplier: Cytiva SH30333.01
Description
Chemical Identifiers
| cGMP |
Specifications
| Powder | |
| Non-sterile | |
| No Glutamine, No Phenol Red, No Poloxamer 188, No HEPES, No Sodium Bicarbonate | |
| 5 L | |
| 2° to 8°C, away from light |
| PF-CHO MPS | |
| None | |
| Gowth of multiple Chinese hamster ovary (CHO) cell clones and production of a variety of recombinant proteins with minimal adaptation | |
| cGMP |
Safety and Handling
ShelfLife : 36 Months