missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva HyClone™ Newborn Calf Serum (U.S.)
Augment cell culture growth with U.S.-sourced HyClone™ Newborn Bovine Calf Serum.
Supplier: Cytiva SH3011803
Description
- Filtered through sequential 100nm filters and sourced from animals that are typically less than 10 days old
Specifications
| Albumin, Iron, Lactate Dehydrogenase, Osmolality, Chloride, Creatinine, Potassium, SGOT, SGPT, Inorganic Phosphorus, TIBC, Alkaline Phosphatase, Total Bilirubin, Total Protein, Blood Urea Nitrogen, Calcium, Chloride, Creatinine, Gamma Globulin, Glucose, IgG, Inorganic Phosphorus | |
| 500 mL | |
| Sequential Triple-filtered, 100 nm | |
| Bluetongue, Bovine Adenovirus, Bovine Parvovirus, Bovine RSV, BVDV, Cytopathogenic Agents, Hemadsorbing Agents, Rabies Virus, Reovirus |
| Newborn Calf Serum (U.S.) | |
| Sterile | |
| None |