missing translation for 'onlineSavingsMsg'
Learn More

Cytiva HyClone™ Newborn Calf Serum (U.S.)

Catalog No. SH3011804
Change view
Click to view available options
Quantity:
500 mL
1000 mL
Processing:
None
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Processing
SH3011804 1000 mL None
SH3011803 500 mL None
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. SH3011804 Supplier Cytiva Supplier No. SH30118.04
NLSU: Get 1 Consumable Product When You Buy 1 Obtenez 1 produit consommable à l’achat d’un produit
View all promotions

Augment cell culture growth with U.S.-sourced HyClone™ Newborn Bovine Calf Serum.

  • Filtered through sequential 100nm filters and sourced from animals that are typically less than 10 days old

Specifications

Additional Assays Albumin, Alkaline Phosphatase, Blood Urea Nitrogen, Calcium, Chloride, Creatinine, Gamma Globulin, Glucose, IgG, Inorganic Phosphorus, Iron, Lactate Dehydrogenase, Osmolality, Percent Saturation, pH, Potassium, SGOT, SGPT, Sodium, TIBC, Total Bilirubin, Total Protein
Description Newborn Calf Serum (U.S.)
Quantity 1000 mL
Sterility Sterile
Sterilization Method Sequential Triple-filtered, 100 nm
Processing None
Tested For Bluetongue, Bovine Adenovirus, Bovine Parvovirus, Bovine RSV, BVDV, Cytopathogenic Agents, Hemadsorbing Agents, Rabies Virus, Reovirus