missing translation for 'onlineSavingsMsg'
Learn More

Cytiva HyClone™ Newborn Bovine Calf Serum, Heat Inactivated

Augment cell culture growth with this cost-effective alternative to Fetal Bovine Serum (FBS)

$278.73

Specifications

Content And Storage Store at or below -10°C
Age <10 days
Country of Origin United States
Form Liquid
Product Type Newborn Calf Bovine Serum
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
SH3011803HI
View Documents Promotion Details
Cytiva
SH30118.03HI
500 mL
Each for $278.73
Only null left
 
Description

Description

  • Produced using careful collection and processing procedures, including venipuncture, to provide the highest quality product available
  • Ensures excellent culture results without fear of contamination
  • HyClone™ maintains complete control of the entire collection and production process for 100% traceability
  • Newborn bovine calf serum undergoes the same careful collection and processing procedures (including venipuncture) used for other bovine serum products
  • Sourced from the United States with complete traceability back to original source
  • Age of animal at time of collection less than 10 days old
  • Low in antibodies and high in growth factors
  • Virus panel testing according to 9 CFR 113.53
Specifications

Specifications

Store at or below -10°C
United States
Newborn Calf Bovine Serum
Sequential-filtered, 100 nm
≤ 50 EU/mL
Yes
HyClone
<10 days
Liquid
Sterile
Heat Inactivated
Augmenting Cell Culture Growth
≤ 25 mg/dL
Frozen
Videos
SDS
Documents

Documents