missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva HyClone™ Newborn Bovine Calf Serum
Description
- Supplement cell culture growth with New Zealand sourced Cytiva HyClone Newborn Bovine Calf Serum, an excellent and cost-effective alternative to fetal bovine serum (FBS).
- Sourced from animals that are typically less than 10 days old
- Traceable
- Conforms to USDA regulations
Specifications
Specifications
| Content And Storage | -10°C or Lower, Once Thawed Store at 2°C to 8°C |
| Country of Origin | New Zealand |
| Product Type | Newborn Calf Bovine Serum |
| Quantity | 25 to 40 Doses |
| Volume (Metric) | 1000 mL |
| For Use With (Application) | Enhance Growth of Cells |