missing translation for 'onlineSavingsMsg'
Learn More

Cytiva HyClone™ Newborn Bovine Calf Serum

Catalog No. SH3412IH254
Change view
Click to view available options
Quantity:
25 to 40 Doses
30 to 45 Doses
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SH3412IH254 25 to 40 Doses
SH3412IH345 30 to 45 Doses
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. SH3412IH254 Supplier Cytiva Supplier No. SH30401.02IH2540
NLSU: Get 1 Consumable Product When You Buy 1 Obtenez 1 produit consommable à l’achat d’un produit
View all promotions

Supplement cell culture growth with New Zealand sourced Cytiva HyClone Newborn Bovine Calf Serum, an excellent and cost-effective alternative to fetal bovine serum (FBS).

  • Supplement cell culture growth with New Zealand sourced Cytiva HyClone Newborn Bovine Calf Serum, an excellent and cost-effective alternative to fetal bovine serum (FBS).
    • Sourced from animals that are typically less than 10 days old
    • Traceable
    • Conforms to USDA regulations

Specifications

Content And Storage -10°C or Lower, Once Thawed Store at 2°C to 8°C
Country of Origin New Zealand
Product Type Newborn Calf Bovine Serum
Quantity 25 to 40 Doses
Volume (Metric) 1000 mL
For Use With (Application) Enhance Growth of Cells