missing translation for 'onlineSavingsMsg'
Learn More

Cytiva HyClone™ Iron-Supplemented Calf Serum, U.S. origin

A cost-effective alternative to Fetal Bovine Serum (FBS). Contains additional iron to boost serum transferrin to physiological levels.

Supplier:  Cytiva SH30072.03

 View more versions of this product

Catalog No. SH3007203

Only null left

Description

Description

HyClone Iron-Supplemented Bovine Calf Serum, US Origin is collected by venipuncture from formula-fed veal calves, which produce exceptionally high levels of transferrin. This serum is carefully supplemented with iron to load serum transferrin levels to physiological levels. After supplementation, the serum contains three to four times as much available iron and transferrin as fetal bovine serum (FBS) or equine serum.

Specifications

Chemical Identifiers

Sterile
Sequential-filtered, 100 nm

Specifications

Insulin, Gamma Globulin, Alkaline Phosphatase, Lactate Dehydrogenase, SGPT, SGOT, pH, Total Protein, Albumin, Blood Urea Nitrogen, Creatinine, Total Bilirubin, Sodium, Potassium, Calcium, Chloride, Phosphorus, Osmolality, Iron, TIBC, Percent Saturation, Glucose
Sera should be stored at -10°C or lower. Once thawed, sera should be stored at 2° to 8°C.
Calf Bovine Serum
Sterile
None
Yes
Manufactured in cGMP (21 CFR 820) compliant and ISO9001 certified facilities
Calf Serum (U.S.), Iron-Supplemented
500 mL
Sequential-filtered, 100 nm
Bluetongue, Bovine Adenovirus, Bovine Parvovirus, Bovine RSV, BVDV, Cytopathogenic Agents, Hemadsorbing Agents, Rabies Virus, Reovirus
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions