missing translation for 'onlineSavingsMsg'
Learn More

Cytiva HyClone™ Iron-Supplemented Calf Serum, U.S. origin

Catalog No. SH3007202HI
Change view
Click to view available options
Quantity:
100 mL
500 mL
1000 mL
Processing:
Heat Inactivated
None
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Processing
SH3007202HI 100 mL Heat Inactivated
SH3007204HI 1000 mL Heat Inactivated
SH3007203 500 mL None
SH3007204 1000 mL None
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. SH3007202HI Supplier Cytiva Supplier No. SH30072.02HI
NLSU: Get 1 Consumable Product When You Buy 1 Obtenez 1 produit consommable à l’achat d’un produit
View all promotions

A cost-effective alternative to Fetal Bovine Serum (FBS). Contains additional iron to boost serum transferrin to physiological levels.

HyClone Iron-Supplemented Bovine Calf Serum, US Origin is collected by venipuncture from formula-fed veal calves, which produce exceptionally high levels of transferrin. This serum is carefully supplemented with iron to load serum transferrin levels to physiological levels. After supplementation, the serum contains three to four times as much available iron and transferrin as fetal bovine serum (FBS) or equine serum.

Specifications

Additional Assays Insulin,Gamma Globulin,Alkaline Phosphatase,Lactate Dehydrogenase,SGPT,SGOT,pH,Albumin,Blood Urea Nitrogen,Creatinine,Total Bilirubin,Sodium,Potassium,Calcium,Chloride,Inorganic Phosphorus,Iron,TIBC,Percent Saturation,Glucose,IgG
Content And Storage Sera should be stored at -10°C or lower. Once thawed, sera should be stored at 2° to 8°C
Product Type Calf Bovine Serum
Quantity 100 mL
Sterility Sterile
Sterilization Method Sequential-filtered, 100 nm
Processing Heat Inactivated
Tested For Bluetongue,Bovine Adenovirus,Bovine Parvovirus,Bovine RSV,BVDV,Cytopathogenic Agents,Hemadsorbing Agents,Rabies Virus,Reovirus
Heat Inactivated Yes