missing translation for 'onlineSavingsMsg'
Learn More

Cytiva HyClone™ Iron-Supplemented Bovine Calf Serum, US Origin

Catalog No. SH204IH2540
Change view
Click to view available options
Quantity:
1000 mL
Processing:
HI, Irr - 25 to 40 doses
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Processing
SH204IH2540 1000 mL HI, Irr - 25 to 40 doses
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SH204IH2540 Supplier Cytiva Supplier No. SH30072.04IH2540
Get 1 Consumable Product When You Buy 1 Obtenez 1 produit consommable à l’achat d’un produit
View all promotions

Contains additional iron that boosts serum transferrin levels to physiological levels

  • Supplemented with iron and growth-promoting factors
  • Age of animal at time of collection approximately 16-24 weeks
  • Virus panel testing according to 9 CFR 113.53
  • Low in antibodies and high in growth factors
  • Sourced in the USA with complete traceability back to original source
  • A cost-effective alternative to FBS

Specifications

Certifications/Compliance Manufactured in cGMP (21 CFR 820) Compliant and ISO9001 Certified Facilities
Age 16 to 22 weeks
Country of Origin United States
Product Type Calf Bovine Serum
Quantity 1000 mL
Sterilization Method Sequential-filtered, 100 nm
Source Formula-Fed Veal Calves
Processing HI, Irr - 25 to 40 doses
Tested For Virus Panel Testing According to 9 CFR 113.53
Endotoxin Concentration <10 EU/mL
For Use With (Application) Supplementing Cell Culture Growth
Heat Inactivated Yes
Hemoglobin Concentration <10 mg/dL
Iron Supplemented Yes
Product Line HyClone
Show More Show Less