missing translation for 'onlineSavingsMsg'
Learn More

Cytiva HyClone™ Insect Cell Screened Characterized Fetal Bovine Serum, US Origin

Catalog No. SH3007103I
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SH3007103I 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SH3007103I Supplier Cytiva Supplier No. SH30071.03I
NLSU: Get 1 Consumable Product When You Buy 1 Obtenez 1 produit consommable à l’achat d’un produit
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Optimizes the performance of insect cells

As Baculovirus Expression Vector Systems (BEVS) increase in popularity, more researchers are culturing insect cells. Insect Cell Screened FBS supports robust growth of insect cells.

Specifications

Color Amber
Content And Storage Store at or Below -10°C
Species Bovine
Age Fetal
Country of Origin United States
Form Liquid
Format Bottle
Specialty Type Insect Cell Screened
Product Type Fetal Bovine Serum
Quantity 500 mL
Sterility Sterile-filtered
Sterilization Method 40 nm filtration
Grade Characterized
Processing Insect Cell Screened
Tested For Bluetongue, Bovine Adenovirus, Bovine Parvovirus, Bovine RSV, BVDV, Cytopathogenic Agents, Hemadsorbing Agents, Rabies Virus, Reovirus
Volume (Metric) 500 mL
Endotoxin Concentration ≤ 10 EU/mL
Heat Inactivated No
Hemoglobin Concentration ≤ 10 mg/dL
Purity or Quality Grade Premium
Product Line HyClone
Osmolality 280 to 360 mOsm/kg
pH 6.7 to 7.6
Protein Content 3 to 4.5 g/dL
Shipping Condition Frozen
Gamma-irradiated No
Show More Show Less