missing translation for 'onlineSavingsMsg'
Learn More

Cytiva HyClone™ HyCell CHO Powder Cell Culture Medium without L-glutamine and with HT

Catalog No. SH3093401
Change view
Click to view available options
Quantity:
1000 mL PETE bottle
5 L
10 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SH3093401 1000 mL PETE bottle
SH3093302 10 L
SH3093301 5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. SH3093401 Supplier Cytiva Supplier No. SH30934.01
NLSU: Get 1 Consumable Product When You Buy 1 Obtenez 1 produit consommable à l’achat d’un produit
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

A versatile, animal derived component free (ADCF) cell culture media.

  • Chemically defined and animal-derived component‐free formulations
  • Designed for high cell yield and recombinant protein production
  • Allow direct or sequential adaptation
  • Designed for large-scale culture applications, including perfusion and fed-batch strategies
  • Developed for Chinese hamster ovary (CHO) cell culture in bioprocessing applications
  • Specifically formulated to provide excellent performance, support high peak cell density, and enhance product yield and quality
  • Can be used for batch, fed-batch, or perfusion applications in multiple cell culture platforms ranging from shake flasks to large-scale bioreactors
  • Specifications

    Form Powder
    Sterility Non-sterile
    With Additives Sodium Bicarbonate, Poloxamer 188, Hypoxanthine, Thymidine
    Without Additives No Phenol Red, No HEPES, No Glutamine
    Quantity 1000 mL PETE bottle
    Content And Storage 2° to 8°C, away from light and moisture