missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva HyClone™ HyCell CHO Powder Cell Culture Medium without L-glutamine and with HT
A versatile, animal derived component free (ADCF) cell culture media.
Supplier: Cytiva SH30933.01
Description
Chemical Identifiers
| Powder | |
| Sodium Bicarbonate, Poloxamer 188, Hypoxanthine, Thymidine | |
| 2° to 8°C, away from light and moisture |
| Non-sterile | |
| No Phenol Red, No HEPES, No Glutamine |
Specifications
| Powder | |
| Sodium Bicarbonate, Poloxamer 188, Hypoxanthine, Thymidine | |
| 5 L |
| Non-sterile | |
| No Phenol Red, No HEPES, No Glutamine | |
| 2° to 8°C, away from light and moisture |
Safety and Handling
ShelfLife : 36 Months