missing translation for 'onlineSavingsMsg'
Learn More

Cytiva HyClone™ HyCell™ CHO Medium, Liquid

Catalog No. SH3094902
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SH3094902 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SH3094902 Supplier Cytiva Supplier No. SH30949.02
NLSU: Get 1 Consumable Product When You Buy 1 Obtenez 1 produit consommable à l’achat d’un produit
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Metabolically designed for high cell yield and recombinant protein production

  • ACDF and chemically-defined cell culture media
  • High-performance growth in multiple culture systems including high-density bioreactors
  • Allows for direct or sequential adaptation
  • Supports multiple CHO clones
  • Component traceability
  • Manufactured in cGMP (21 CFR 820) compliant and ISO9001 certified facilities
  • Sourced from animals that are typically less than 10 days old

Specifications

Form Liquid
Product Type CHO Medium
For Use With (Application) Growth Promotion
Product Line HyCell
Quantity 1 L
Certifications/Compliance Manufactured in cGMP (21 CFR 820) Compliant and ISO9001 Certified Facilities
Endotoxin Level <10 EU/mL
Format Bottle
Osmolality 270 to 330 mOsm/kg
pH 7.0 to 7.4
Color Clear Yellow
Description With Sodium Bicarbonate/Poloxamer 188, Without HT/L-Glutamine
Show More Show Less