missing translation for 'onlineSavingsMsg'
Learn More

Cytiva HyClone™ Human Mesenchymal Stem Cell Screened Characterized Fetal Bovine Serum, US Origin

Catalog No. SH3007103M
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SH3007103M 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SH3007103M Supplier Cytiva Supplier No. SH30071.03M
NLSU: Get 1 Consumable Product When You Buy 1 Obtenez 1 produit consommable à l’achat d’un produit
View all promotions

Designed for human mesenchymal stem cells

Human Mesenchymal Stem Cell Screened FBS supports the growth of undifferentiated human mesenchymal stem cells (hMSC). During screening, hMSC are observed for evidence of nutritional deficiency, cytotoxicity, or morphological aberrations.

Specifications

Country of Origin United States
Form Liquid
Quantity 500 mL
Sterilization Method 40 nm filtration
Endotoxin Concentration ≤ 10 EU/mL
Heat Inactivated No
Hemoglobin Concentration ≤ 10 mg/dL