missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva HyClone™ FetalClone™ III Serum (U.S.)
Designed for fibroblast applications, supplement cell culture growth with U.S.-sourced HyClone™ FetalClone™ III Serum, the most widely applicable fetal bovine serum (FBS) alternative.
Supplier: Cytiva SH30109.02HI
Description
- Economical alternative to FBS
- Performance comparable with FBS
- High lot-to-lot consistency
- Low endotoxin and hemoglobin levels
Chemical Identifiers
| Manufactured in cGMP (21 CFR 820) compliant and ISO9001 certified facilities | |
| United States | |
| Sterile | |
| ≤ 10 EU/mL | |
| Typically less than 2 μg/mL | |
| No |
| Sera should be stored at -10°C or lower. Once thawed, sera should be stored at 2 to 8°C. | |
| FBS Replacement Serum | |
| Sequential-filtered, 100 nm | |
| ≤ 20 mg/dL | |
| HyClone | |
| USDA Inspected |
Specifications
| Gamma Globulin,Alkaline Phosphatase,Lactate Dehydrogenase,SGPT,SGOT,pH,Total Protein,Albumin,Blood Urea Nitrogen,Creatinine,Total Bilirubin,Sodium,Potassium,Calcium,Chloride,Inorganic Phosphorus,Osmolality,Iron,TIBC,Percent Iron Saturation,Glucose,IgG | |
| Sera should be stored at -10°C or lower. Once thawed, sera should be stored at 2 to 8°C. | |
| FBS Replacement Serum | |
| Sterile | |
| Heat Inactivated | |
| ≤ 10 EU/mL | |
| ≤ 20 mg/dL | |
| HyClone | |
| USDA Inspected |
| Manufactured in cGMP (21 CFR 820) compliant and ISO9001 certified facilities | |
| United States | |
| 100 mL | |
| Sequential-filtered, 100 nm | |
| Bluetongue, Bovine Adenovirus, Bovine Parvovirus, Bovine RSV, BVDV, Cytopathogenic Agents, Hemadsorbing Agents, Rabies Virus, Reovirus | |
| Yes | |
| Typically less than 2 μg/mL | |
| No |