missing translation for 'onlineSavingsMsg'
Learn More

Cytiva HyClone™ Fetal Clone I Serum

Catalog No. p-8013213
Change view
Click to view available options
Quantity:
500 mL
Processing:
Heat Inactivated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Processing
SH3008003HI 500 mL Heat Inactivated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SH3008003HI Supplier Cytiva Supplier No. SH30080.03HI
Only null left

HyClone™ FetalClone™ I is a fetal bovine serum alternative optimised for the growth of hybridomas with comparable IgG levels to those found in FBS.

Specifications

Additional Assays Gamma Globulin, Alkaline Phosphatase, Lactate Dehydrogenase, SGPT, SGOT, pH, Total Protein, Albumin, Blood Urea Nitrogen, Creatinine, Total Bilirubin, Sodium, Potassium, Calcium, Chloride, Inorganic Phosphorus, Osmolality, Iron, TIBC, Percent Iron Saturation, Glucose, IgG
Certifications/Compliance Manufactured in cGMP (21 CFR 820) compliant and ISO9001 certified facilities
Content And Storage Sera should be stored at -10°C or lower. Once thawed, sera should be stored at 2° to 8°C.
Description FetalClone I Serum (U.S.) Heat Inactivated
Country of Origin United States
Product Type FBS Replacement Serum
Quantity 500 mL
Sterility Sterile
Sterilization Method Sequential-filtered, 100 nm
Processing Heat Inactivated
Tested For Bluetongue, Bovine Adenovirus, Bovine Parvovirus, Bovine RSV, BVDV, Cytopathogenic Agents, Hemadsorbing Agents, Rabies Virus, Reovirus
Endotoxin Concentration ≤ 10 EU/mL
Heat Inactivated Yes
Hemoglobin Concentration ≤ 20 mg/dL
IgG levels Typically less than 2 μg/mL
Mycoplasma Detected No
USDA Safety Testing Included USDA Inspected
Show More Show Less