missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva HyClone™ Fetal Bovine Serum (U.S.), Dialyzed
For applications that need serum without low molecular weight components.
Supplier: Cytiva SH3007903IH2540
Description
Augment growth of cells with U.S.-sourced HyClone™ Dialyzed Fetal Bovine Serum (FBS), depleted of small molecules (less than 10,000 MW) and filtered through sequential 100nm (0.1μm) pore-size rated filters.
Specifications
| 25 Dihydroxy Vitamin D, Albumin, Alkaline Phosphatase, B1 | |
| Sera should be stored at -10°C or lower. Once thawed, sera should be stored at 2°C to 8°C. | |
| Dialyzed | |
| Sterile | |
| Specialty filtered | |
| Bluetongue, Bovine Adenovirus, Bovine Parvovirus, Bovine RSV, BVDV | |
| Yes | |
| No |
| EMEA Certified | |
| United States | |
| 500 mL | |
| Sequential-filtered, 100 nm | |
| HI, Irr - 25 to 40 doses | |
| ≤ 10 EU/mL | |
| ≤ 10 mg/dL | |
| USDA Inspected |