missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva HyClone™ Fetal Bovine Serum (U.S.), Dialyzed
For applications that need serum without low molecular weight components.
Supplier: Cytiva SH3007901HI
Description
Augment growth of cells with U.S.-sourced HyClone™ Dialyzed Fetal Bovine Serum (FBS), depleted of small molecules (less than 10,000 MW) and filtered through sequential 100nm (0.1μm) pore-size rated filters.
Specifications
| 1, Calcium, Carotene, Chloride, Cholesterol, Copper, Creatinine, Folate, Gamma Globulin, Glucose, HDL, 25 Dihydroxy Vitamin D, IgG, Inorganic Phosphorus, Insulin, Iron, Lactate Dehydrogenase, LDL, Magnesium, Niacin, Osmolality, Percent Saturation, Albumin, pH, Phospholipids, Potassium, Progesterone, Retinol, Selenium, SGOT, SGPT, Sodium, TIBC, Alkaline Phosphatase, Tocopherol, Total Bilirubin, Total Protein, Triglycerides, Vitamin C, B1, B12, B2, B6, Biotin, Blood Urea Nitrogen | |
| Dialyzed Fetal Bovine Serum, US Origin, Heat Inactivated | |
| Dialyzed | |
| 50 mL | |
| Sequential-filtered, 100 nm | |
| Heat Inactivated | |
| ≤ 10 EU/mL | |
| ≤ 10 mg/dL | |
| USDA Inspected |
| Sera should be stored at -10°C or lower. Once thawed, sera should be stored at 2° to 8°C. | |
| United States | |
| Fetal Bovine Serum | |
| Sterile | |
| Specialty filtered | |
| Bluetongue, Bovine Adenovirus, Bovine Parvovirus, Bovine RSV, BVDV, Cytopathogenic Agents, Hemadsorbing Agents, Rabies Virus, Reovirus | |
| Yes | |
| No |