missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva Hyclone™ Defined Fetal Bovine Serum (U.S.)
Used as serum-supplement for the in vitro cell culture of eukaryotic cells
Supplier: Cytiva SH3007003
Description
- Maximize cell culture growth
- Tested for endotoxin, hemoglobin, and additional biochemical assays
- Filtered through serial 40 nm pore-size rated filters
Specifications
| 1, Calcium, Albumin, Chloride, Cholesterol, B12, B2, Folate, Biotin, Glucose, HDL, 25 Dihydroxy Vitamin D, IgG, Inorganic Phosphorus, Insulin, Iron, Lactate Dehydrogenase, LDL, Magnesium, Niacin, Osmolality, Percent Saturation, Albumin, pH, Phospholipids, Potassium, Progesterone, Retinol, Selenium, SGOT, SGPT, Sodium, TIBC, Alkaline Phosphatase, Tocopherol, Total Bilirubin, Total Protein, Triglycerides, Vitamin C, B1, B12, B2, B6, Biotin, Blood Urea Nitrogen | |
| Sera should be stored at -10°C or lower. Once thawed, sera should be stored at 2 to 8°C. | |
| Fetal Bovine Serum | |
| Sterile | |
| Defined | |
| Bluetongue, Bovine Adenovirus, Bovine Parvovirus, Bovine RSV, BVDV, Cytopathogenic Agents, Hemadsorbing Agents, Rabies Virus, Reovirus | |
| No | |
| HyClone | |
| USDA Inspected |
| Manufactured in cGMP (21 CFR 820) compliant and ISO9001 certified facilities | |
| United States | |
| 500 mL | |
| Sequential-filtered, 40 nm | |
| None | |
| ≤ 10 EU/mL | |
| ≤ 10 mg/dL | |
| No |