missing translation for 'onlineSavingsMsg'
Learn More

Cytiva HyClone™ Fetal Bovine Serum (Canada), Characterized

Catalog No. SH3039603HI
Change view
Click to view available options
Quantity:
100 mL
500 mL
Processing:
Gamma Irr, 25 to 40 doses
Heat Inactivated
None
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Processing
SH3039603HI 500 mL Heat Inactivated
SH3039602 100 mL None
SH3039603 500 mL None
SH303960340 500 mL Gamma Irr, 25 to 40 doses
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. SH3039603HI Supplier Cytiva Supplier No. SH30396.03HI
NLSU: Get 1 Consumable Product When You Buy 1 Obtenez 1 produit consommable à l’achat d’un produit
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Cytiva™ Characterized Fetal Bovine Serum, Canadian Origin enhances the growth of cells with Canadian sourced HyClone Characterized Fetal Bovine Serum.

HyClone™ Characterized Fetal Bovine Serum is filtered using 100nm (0.1μm) pore-size filters and processed using True Pool processing technology to ensure consistency in the quality of serum. HyClone™ Characterized FBS is widely used by professionals requiring an extensive biochemical profile and full traceability compliant with current Good Manufacturing Practices (cGMP).

Characterized FBS is ≤ 25EU/mL Endotoxin and ≤ 25mg/mL Hemoglobin.

  • Characterized Fetal Bovine Serum (FBS), Canadian origin, is filtered using 100 nm (0.1 μm) pore size-rated filters and processed using true pool technology to ensure consistency in the quality of the serum (Fig 1). Characterized FBS is widely used by professionals requiring an extensive biochemical profile and full traceability back to the original source. Characterized FBS, Canadian Origin is compliant with current good manufacturing practices (cGMP).
  • Characterized FBS is filtered through three sequential 100 nm (0.1 μm) pore size-rated filters. Before dispensing, each lot of serum is pooled to ensure uniformity and consistency between bottles (true pool technology). Characterized FBS is assayed for gamma globulin, alkaline phosphatase, lactate dehydrogenase, glutamic pyruvic transaminase (SBPT), glutamic oxaloacetic transaminase (SGOT), pH, total protein, albumin, blood urea nitrogen, creatinine, total bilirubin, sodium, potassium, calcium, chloride, inorganic phosphorus, osmolality, iron, total iron binding capacity (TIBC), percent iron saturation, glucose, and IgG. Included assays are subject to change without notice.

Specifications

Additional Assays Albumin, Alkaline Phosphatase, Blood Urea Nitrogen, Calcium, Chloride, Creatinine, Gamma Globulin, Glucose, IgG, Inorganic Phosphorus, Iron, Lactate Dehydrogenase, Osmolality, Percent Saturation, pH, Potassium, SGOT, SGPT, Sodium, TIBC, Total Bilirubin, Total Protein
Certifications/Compliance EMEA Certified
Content And Storage Sera should be stored at -10°C or lower. Once thawed, sera should be stored at 2° to 8°C.
Description Characterized Fetal Bovine Serum, Canadian Origin, Heat Inactivated
Country of Origin Canada
Quantity 500 mL
Sterility Sterile
Sterilization Method Sequential-filtered, 100 nm
Grade Characterized
Processing Heat Inactivated
Tested For Bluetongue, Bovine Adenovirus, Bovine Parvovirus, Bovine RSV, BVDV, Cytopathogenic Agents, Hemadsorbing Agents, Rabies Virus, Reovirus
Endotoxin Concentration ≤ 25 EU/mL
Heat Inactivated Yes
Hemoglobin Concentration ≤ 25 mg/dL
Mycoplasma Detected No
USDA Safety Testing Included USDA Inspected
Show More Show Less