missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva Hyclone™ Characterized Fetal Bovine Serum (Australia)
Characterized FBS irradiated is comparable to the highest quality FBS from other suppliers, but is lower priced than defined FBS.
$1,219.79
Specifications
| Certifications/Compliance | AQIS |
|---|---|
| Content And Storage | Sera should be stored at -10°C or lower. Once thawed, sera should be stored at 2 to 8°C. |
| Country of Origin | Australia |
| Product Type | Fetal Bovine Serum |
| Sterility | Sterile |
| Catalog Number | Mfr. No. | Quantity | Processing | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Processing | Price | Quantity & Availability | ||||
|
SH3008403HI
|
Cytiva
SH30084.03HI |
500 mL | Heat Inactivated |
Each for $1,219.79
|
|
||||
Description
- Filtered through 3 sequential 0.1 μm pore size filters
Specifications
| AQIS | |
| Australia | |
| Sterile | |
| USDA Inspected |
| Sera should be stored at -10°C or lower. Once thawed, sera should be stored at 2 to 8°C. | |
| Fetal Bovine Serum | |
| HyClone |