Learn More
Cytiva HyClone™ ES Screened Characterized Fetal Bovine Serum, US Origin
Optimized for the culture of murine embryonic stem cells
Supplier: Cytiva SH3007103E
Description
ES Screened FBS supports the growth of undifferentiated murine embryonic stem cells. The screening includes plating efficiency, colony morphology, and toxicity tests.Specifications
| United States | |
| ES cell screened | |
| 40 nm filtration | |
| No |
| Liquid | |
| 500 mL | |
| ≤ 10 EU/mL | |
| ≤ 10 mg/dL |