missing translation for 'onlineSavingsMsg'
Learn More

Cytiva HyClone™ ES Screened Characterized Fetal Bovine Serum, US Origin

Catalog No. SH3007103EH
Change view
Click to view available options
Quantity:
100 mL
500 mL
Heat Inactivated:
No
Yes
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Heat Inactivated
SH3007103EH 500 mL Yes
SH3007102E 100 mL No
SH3007102EH 100 mL Yes
SH3007103E 500 mL No
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. SH3007103EH Supplier Cytiva Supplier No. SH30071.03EH
Get 1 Consumable Product When You Buy 1 Obtenez 1 produit consommable à l’achat d’un produit
View all promotions

Optimized for the culture of murine embryonic stem cells

ES Screened FBS supports the growth of undifferentiated murine embryonic stem cells. The screening includes plating efficiency, colony morphology, and toxicity tests.

Specifications

Country of Origin United States
Form Liquid
Quantity 500 mL
Sterilization Method 40 nm filtration
Processing Heat Inactivated
Endotoxin Concentration ≤ 10 EU/mL
Heat Inactivated Yes
Hemoglobin Concentration ≤ 10 mg/dL