missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva HyClone™ Dulbecco's Phosphate Buffered Saline, Solution
Buffer cell culture with PBS in solution.
Supplier: Cytiva SH30264.FS
Description
- Format: Liquid
Chemical Identifiers
| Manufactured under cGMP and ISO 9001 certified facilities | |
| Liquid | |
| Sterile | |
| ≤0.025EU/mL | |
| 276±10mOsm/kg H2O | |
| RUO |
| 15° to 30°C | |
| Dulbecco's Phosphate Buffered Saline | |
| 0.1 μm Sterile-filtered | |
| Serves as an irrigating, transporting or diluting fluid while maintaining cell tonicity and viability for a limited period of time | |
| 7 to 7.6 |
Specifications
| Manufactured under cGMP and ISO 9001 certified facilities | |
| HyClone Dulbecco's Phosphate Buffered Salines (DPBS) With Calcium, Magnesium, without Phenol Red | |
| Liquid | |
| Sterile | |
| Calcium, Magnesium | |
| ≤0.025EU/mL | |
| 276±10mOsm/kg H2O | |
| 6 x 500 mL |
| 15° to 30°C | |
| 1 X | |
| Dulbecco's Phosphate Buffered Saline | |
| 0.1 μm Sterile-filtered | |
| No Phenol Red | |
| Serves as an irrigating, transporting or diluting fluid while maintaining cell tonicity and viability for a limited period of time | |
| 7 to 7.6 | |
| RUO |
Safety and Handling
ShelfLife : 24 Months