missing translation for 'onlineSavingsMsg'
Learn More

Cytiva HyClone™ Dulbecco's Phosphate Buffered Saline, Solution

Buffer cell culture with PBS in solution.

Supplier:  Cytiva SH30264.01

 View more versions of this product

Catalog No. SH3026401

Only null left

Description

Description

  • Format: Liquid
Specifications

Chemical Identifiers

Manufactured under cGMP and ISO 9001 certified facilities
Liquid
Sterile
≤0.025EU/mL
276±10mOsm/kg H2O
RUO
15° to 30°C
Dulbecco's Phosphate Buffered Saline
0.1 μm Sterile-filtered
Serves as an irrigating, transporting or diluting fluid while maintaining cell tonicity and viability for a limited period of time
7 to 7.6

Specifications

Manufactured under cGMP and ISO 9001 certified facilities
HyClone Dulbecco's Phosphate Buffered Salines (DPBS) With Calcium, Magnesium, without Phenol Red
Liquid
Sterile
Calcium, Magnesium
≤0.025EU/mL
276±10mOsm/kg H2O
500 mL
15° to 30°C
1 X
Dulbecco's Phosphate Buffered Saline
0.1 μm Sterile-filtered
No Phenol Red
Serves as an irrigating, transporting or diluting fluid while maintaining cell tonicity and viability for a limited period of time
7 to 7.6
RUO
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

ShelfLife : 24 Months

SDS
Documents

Documents

Promotions

Promotions