missing translation for 'onlineSavingsMsg'
Learn More

Cytiva HyClone™ Dulbecco's Phosphate Buffered Saline, Powder

Catalog No. SH3001304
Change view
Click to view available options
Quantity:
10 L
2 x 5 L
50 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SH3001304 50 L
SH3001302 2 x 5 L
SH3001303 10 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. SH3001304 Supplier Cytiva Supplier No. SH30013.04
NLSU: Get 1 Consumable Product When You Buy 1 Obtenez 1 produit consommable à l’achat d’un produit
View all promotions

Buffer cell culture with HyClone™ Dulbecco's Phosphate Buffered Saline (PBS), Powder.

Specifications

Certifications/Compliance Manufactured in cGMP (21 CFR 820) compliant and ISO9001 certified facilities
Content And Storage 2° to 8° C
Form Powder
Sterility Non-sterile
Without Additives No Calcium, No Magnesium
Quantity 50 L