missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva HyClone™ DPM-PF-CHO Powdered Media
Description
- Designed for high cell yield and recombinant protein production
- Allow for direct or sequential adaptation
- Support growth in multiple cell culture systems
- Manufactured from traceable components according to cGMP (21 CFR 820) guidelines
Specifications
Specifications
| Form | Powder |
| Product Type | DPM-PF-CHO Base |
| Sterility | Non-sterile |
| Without Additives | No Glutamine, No Phenol Red, No Poloxamer 188 |
| Quantity | 5 L |
| Content And Storage | 2 to 8°C, away from light and moisture |