missing translation for 'onlineSavingsMsg'
Learn More

Cytiva HyClone™ DPM-PF-CHO Powdered Media

Catalog No. SH3033501
Change view
Click to view available options
Product Type:
DPM-PF-CHO Base
DPM-PF-CHO Main
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
SH3033501 DPM-PF-CHO Base
SH3033401 DPM-PF-CHO Main
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. SH3033501 Supplier Cytiva Supplier No. SH30335.01
NLSU: Get 1 Consumable Product When You Buy 1 Obtenez 1 produit consommable à l’achat d’un produit
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Powder mediums (Main/Base) used to create PF-CHO MPS media

  • Designed for high cell yield and recombinant protein production
  • Allow for direct or sequential adaptation
  • Support growth in multiple cell culture systems
  • Manufactured from traceable components according to cGMP (21 CFR 820) guidelines

Specifications

Form Powder
Product Type DPM-PF-CHO Base
Sterility Non-sterile
Without Additives No Glutamine, No Phenol Red, No Poloxamer 188
Quantity 5 L
Content And Storage 2 to 8°C, away from light and moisture