missing translation for 'onlineSavingsMsg'
Learn More

Cytiva HyClone™ Donor Equine Serum, US Origin, Irradiated 30-45 kGy

Catalog No. SH3744IR345
Change view
Click to view available options
Quantity:
1000 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SH3744IR345 1000 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SH3744IR345 Supplier Cytiva Supplier No. SH30074.04IR3045
NLSU: Get 1 Consumable Product When You Buy 1 Obtenez 1 produit consommable à l’achat d’un produit
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Complete traceability back to the original source.

  • Tested for equine infectious anemia
  • Complete traceability back to original source
  • Virus panel testing according to 9 CFR 113.53

Specifications

Content And Storage Sera should be stored at -10°C or lower. Once thawed, sera should be stored at 2°C to 8°C.
Species Equine
Age Adult
Country of Origin United States
Form Frozen
Format Bottle
Product Type Equine Serum
Quantity 1000 mL
Serum Treatment Gamma-irradiated
Sterility Sterile-filtered
Sterilization Method Irradiated
Endotoxin Concentration ≤ 10 EU/mL
Hemoglobin Concentration ≤ 10 mg/dL
Mycoplasma Detected Not Detected
Gamma-irradiated Yes
Show More Show Less