missing translation for 'onlineSavingsMsg'
Learn More

Cytiva HyClone™ Donor Equine Serum (U.S. origin)

Catalog No. SH3007402HI
Change view
Click to view available options
Quantity:
100 mL
Processing:
Heat Inactivated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Processing
SH3007402HI 100 mL Heat Inactivated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SH3007402HI Supplier Cytiva Supplier No. SH30074.02HI
NLSU: Get 1 Consumable Product When You Buy 1 Obtenez 1 produit consommable à l’achat d’un produit
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Supplement cell culture growth with U.S. sourced, collected from selected donor herds that receive regular veterinary inspection and care. Complete traceability back to the original source.

  • Donor herds are fed an enriched diet to ensure proper nutrition and are fasted prior to bleeding to reduce lipid concentration
  • The horses are bled in an enclosed facility that is specially equipped for aseptic collection
  • Coggins tests for equine infectious anemia are routinely performed on each horse
  • Before dispensing, each lot of serum is pooled using true pool technology to ensure uniformity and consistency between bottles
  • Specifications

    Additional Assays Insulin,Gamma Globulin,Alkaline Phosphatase,Lactate Dehydrogenase,SGPT,SGOT,pH,Albumin,Blood Urea Nitrogen,Creatinine,Total Bilirubin,Sodium,Potassium,Calcium,Chloride,Inorganic Phosphorus,Iron,TIBC,Percent Saturation,Glucose,IgG
    Content And Storage Store at or Below -10°C
    Species Equine
    Age Adult
    Country of Origin United States
    Form Frozen
    Format Bottle
    Product Type Equine Serum
    Quantity 100 mL
    Serum Treatment Heat-inactivated
    Sterility Sterile-filtered
    Sterilization Method Sequential-filtered, 100 nm
    Processing Heat Inactivated
    Tested For Equine Herpesvirus, Equine Viral Arteritis, Bovine Virus Diarrhea, Rabies, Reovirus, Cytopathogenic Agnets (IBR), Hemabsorbing Agent (PI3), Equine Infectious Anemia
    Heat Inactivated Yes
    Product Line HyClone
    Shipping Condition Frozen
    Show More Show Less