missing translation for 'onlineSavingsMsg'
Learn More

Cytiva HyClone™ Cosmic Calf™ Serum (U.S.)

Catalog No. SH3008702
Change view
Click to view available options
Quantity:
100 mL
500 mL
1000 mL
Processing:
Heat Inactivated
None
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Processing
SH3008702 100 mL None
SH3008703 500 mL None
SH3008704 1000 mL None
SH3008703HI 500 mL Heat Inactivated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. SH3008702 Supplier Cytiva Supplier No. SH30087.02
NLSU: Get 1 Consumable Product When You Buy 1 Obtenez 1 produit consommable à l’achat d’un produit
View all promotions

Contains high quality iron-supplemented bovine calf serum with added growth promoting factors.

  • Age at time of collection: typically 16 - 22 weeks
  • Iron-supplemented with additional growth promoting components

Specifications

Additional Assays Albumin, Alkaline Phosphatase, Blood Urea Nitrogen, Calcium, Chloride, Creatinine, Gamma Globulin, Glucose, IgG, Insulin, Iron, Lactate Dehydrogenase, Osmolality, Percent Saturation, pH, Phosphorus, Potassium, SGOT, SGPT, Sodium, TIBC, Total Bilirubin, Total Protein
Certifications/Compliance Manufactured in cGMP (21 CFR 820) compliant and ISO9001 certified facilities
Content And Storage Sera should be stored at -10°C or lower. Once thawed, sera should be stored at 2° to 8°C.
Description Calf Serum; Cosmic Calf; 100 mL
Additive Iron and naturally derived components
Product Type Calf Bovine Serum
Quantity 100 mL
Sterility Sterile
Sterilization Method Sequential-filtered, 100 nm, Gamma-irradiated
Processing None
Tested For Bluetongue, Bovine Adenovirus, Bovine Parvovirus, Bovine RSV, BVDV, Cytopathogenic Agents, Hemadsorbing Agents, Rabies Virus, Reovirus
Show More Show Less