missing translation for 'onlineSavingsMsg'
Learn More

Cytiva HyClone™ Characterized Fetal Bovine Serum (U.S. Origin)

Catalog No. SH3007101
Change view
Click to view available options
Processing:
Heat Inactivated
None
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Processing
SH3007101 None
SH3007101HI Heat Inactivated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. SH3007101 Supplier Cytiva Supplier No. SH30071.01
NLSU: Get 1 Consumable Product When You Buy 1 Obtenez 1 produit consommable à l’achat d’un produit
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Enhance growth of cells. Tested for endotoxin, hemoglobin, and additional biochemical assays.

  • Provides important proteins, hormones, growth factors, metabolites, and nutrients for your cell culture process
  •  Produced using true pool technology, which minimizes bottle-to-bottle variability
  • Assayed for gamma globulin, alkaline phosphatase, lactate dehydrogenase, glutamic pyruvic transaminase, glutamic oxaloacetic transaminase, pH, total protein, albumin, blood urea nitrogen, creatinine, total bilirubin, sodium, potassium, calcium, chloride, inorganic phosphorus, osmolality, iron, total iron binding capacity (TIBC), percent iron saturation, glucose, and IgG
  • U.S. sourced
  • Specifications

    Additional Assays Lactate Dehydrogenase,SGPT,SGOT,pH,Total Protein,Albumin,Blood Urea Nitrogen,Creatinine,Total Bilirubin,Sodium,Potassium,Calcium,Chloride,Inorganic Phosphorus,Osmolality,Iron,TIBC,Percent Saturation,Glucose,IgG,Gamma Globulin
    Certifications/Compliance Manufactured in cGMP (21 CFR 820) compliant and ISO9001 certified facilities
    Content And Storage Sera should be stored at -10°C or lower. Once thawed, sera should be stored at 2° to 8°C.
    Country of Origin United States
    Quantity 50 mL
    Sterility Sterile
    Sterilization Method Sequential Triple-filtered, 100 nm
    Grade Characterized
    Processing None
    Tested For Bluetongue, Bovine Adenovirus, Bovine Parvovirus, Bovine RSV, BVDV, Cytopathogenic Agents, Hemadsorbing Agents, Rabies Virus, Reovirus
    Endotoxin Concentration ≤ 25 EU/mL
    Heat Inactivated No
    Hemoglobin Concentration ≤ 25 mg/dL
    Mycoplasma Detected No
    USDA Safety Testing Included USDA Inspected
    Show More Show Less