missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva HyClone™ Characterized Fetal Bovine Serum, Australian Origin
Description
- Meets the requirements of most users
- Filtered through triple 100 nm pore size-rated filters
- Standard endotoxin and hemoglobin specifications
- Subjected to moderate biochemical analyses
Specifications
Specifications
| Certifications/Compliance | Certificate of Suitability No. R1-CEP 2000-384 |
| Country of Origin | Australia |
| Form | Serum |
| Product Type | Fetal Bovine Serum |
| Quantity | 500 mL |
| Sterilization Method | Sequential Triple-filtered, 100 nm |
| Grade | Characterized |
| Processing | HI, Irr - 25 to 40 doses |
| Endotoxin Concentration | ≤ 25 EU/mL |
| Hemoglobin Concentration | ≤ 25 mg/dL |
| Show More |