missing translation for 'onlineSavingsMsg'
Learn More

Cytiva HyClone™ Cell Boost™ 7b Supplement

Catalog No. SH3102701
Change view
Click to view available options
Quantity:
500 mL
Form:
Powder
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Form
SH3102701 500 mL Powder
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SH3102701 Supplier Cytiva Supplier No. SH31027.01
NLSU: Get 1 Consumable Product When You Buy 1 Obtenez 1 produit consommable à l’achat d’un produit
View all promotions

A concentrated amino acid supplement, in a ready to use powder format, for high-yield protein production in CHO fed-batch processes

  • Improve productivity and cell yield in fed-batch cultures by supplementing the cell culture medium to replace nutrients depleted during cell culture
  • A concentrated solution of amino acids with an alkaline pH
  • Cell Boost 7a and 7b are concentrated cell culture supplements designed to be used together to enhance protein production.
  • Designed to support modern manufacturing processes
  • Chemically defined (CD), animal-derived component-free (ADCF), and does not contain any growth factors (such as insulin), peptides, hydrolysates, phenol red, or 2-mercaptoethanol
  • Intended for use in combination with HyClone ActiPro and ActiSM media as part of the ActiPro cell culture system
  • Formulated to provide high yields of recombinant proteins in processes when using Chinese hamster ovary (CHO) cell lines
  • Manufactured to meet cGMP manufacturing standards and QC specifications

Specifications

Content And Storage Store 2°C to 8°C, Away From Light and Moisture
Description Without: L-glutamine, poloxamer 188, and insulin
Form Powder
Product Type Amino Acid Supplement
Sterility Non-sterile
For Use With (Application) High-Yield Protein Production in CHO Fed-Batch Processes
Product Line HyClone
Quantity 500 mL