missing translation for 'onlineSavingsMsg'
Learn More

Cytiva HyClone™ Cell Boost™ 3 Supplement

Catalog No. SH3111502
Change view
Click to view available options
Quantity:
1000 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SH3111502 1000 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SH3111502 Supplier Cytiva Supplier No. SH31115.02
NLSU: Get 1 Consumable Product When You Buy 1 Obtenez 1 produit consommable à l’achat d’un produit
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Fed-batch supplement with amino acids, vitamins, glucose, and trace elements for hybridoma and myeloma cells

  • Chemically Defined (CD), Animal-Derived Component-Free (ADCF), and Protein-Free
  • Supports fed-batch cultures with amino acids, vitamins, glucose, and trace elements
  • Developed for recombinant protein production with various cell lines including hybridoma and myeloma cell lines
  • Designed to support modern cell culture manufacturing processes in research and bioprocessing applications
  • Manufactured to meet cGMP manufacturing standards and QC specifications
  • Available as a ready-to-use liquid feed solution

Specifications

Cell Type Hybridomas, Myelomas
Form Liquid
Product Type Cell Boost Supplement
Format Bottle
For Use With (Application) Improving Productivity and Cell Growth in Fed-Batch Cultures
Product Line HyClone
Quantity 1000 mL