missing translation for 'onlineSavingsMsg'
Learn More

Cytiva HyClone™ CDM4HEK293 Powdered Media

Catalog No. SH3085901
Change view
Click to view available options
Quantity:
5 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SH3085901 5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SH3085901 Supplier Cytiva Supplier No. SH30859.01
NLSU: Get 1 Consumable Product When You Buy 1 Obtenez 1 produit consommable à l’achat d’un produit
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Promotes adenovirus and recombinant protein production in HEK 293 suspension cell culture.

  • Optimized for high cell growth and high recombinant protein and vector production with human embryonic kidney (HEK) 293 cells
  • Chemically defined, protein-free and animal-derived component-free medium available in liquid and powder form
  • Designed for small- and large-scale biomanufacturing processes
  • Manufactured in cGMP (21 CFR 820) compliant and ISO9001 certified facilities
  • Supports direct or sequential adaptation

Specifications

Content And Storage 2 to 8°C, away from light and moisture
Form Powder
Sterility Non-sterile
Without Additives No Glutamine, No HEPES, No Phenol Red, No Sodium Bicarbonate
Quantity 5 L