missing translation for 'onlineSavingsMsg'
Learn More

Cytiva HyClone™ CDM4CHO™ Media, Liquid

Catalog No. SH3055802
Change view
Click to view available options
Quantity:
1000 mL bottle
1000 mL
With Additives:
Glutamine, Sodium Bicarbonate, Poloxamer 188
Sodium Bicarbonate, Poloxamer 188
Without Additives:
No Glutamine, No Phenol Red, No HEPES, No Protein
No Phenol Red, No HEPES, No Protein
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity With Additives Without Additives
SH3055802 1000 mL Sodium Bicarbonate, Poloxamer 188 No Glutamine, No Phenol Red, No HEPES, No Protein
SH3055702 1000 mL bottle Glutamine, Sodium Bicarbonate, Poloxamer 188 No Phenol Red, No HEPES, No Protein
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. SH3055802 Supplier Cytiva Supplier No. SH30558.02
NLSU: Get 1 Consumable Product When You Buy 1 Obtenez 1 produit consommable à l’achat d’un produit
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Support growth of CHO cell culture. HyClone™ CDM4CHO™ Media, Liquid is a serum-free, chemically defined media.

  • Format: Liquid

Specifications

Form Liquid
Product Type CDM4CHO Liquid Medium
Sterilization Method 0.2 μm Sterile-filtered
With Additives Sodium Bicarbonate, Poloxamer 188
Without Additives No Glutamine, No Phenol Red, No HEPES, No Protein
Quantity 1000 mL
Certifications/Compliance cGMP
Content And Storage 2° to 8°C, away from light
Description CDM4CHO media