missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva HyClone™ CDM4CHO™ Media, Liquid
Support growth of CHO cell culture. HyClone™ CDM4CHO™ Media, Liquid is a serum-free, chemically defined media.
Supplier: Cytiva SH30557.02
Description
- Format: Liquid
Chemical Identifiers
| Liquid | |
| cGMP | |
| CDM4CHO media |
Specifications
| Liquid | |
| 0.2 μm Sterile-filtered | |
| No Phenol Red, No HEPES, No Protein | |
| cGMP | |
| CDM4CHO media |
| CDM4CHO Liquid Medium | |
| Glutamine, Sodium Bicarbonate, Poloxamer 188 | |
| 1000 mL bottle | |
| 2° to 8°C, away from light |
Safety and Handling
ShelfLife : 12 Months