missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva HyClone™ Bovine Growth Serum (U.S.)
Support cell culture growth with U.S.-sourced HyClone™ Bovine Growth Serum, a superior performing fetal bovine serum (FBS) replacement, consisting of bovine calf serum supplemented with chemically-defined components.
Supplier: Cytiva SH3054103
Description
- Filtered through sequential 100nm (0.1μm) pore-size rated filters
Specifications
| Albumin, Alkaline Phosphatase, Blood Urea Nitrogen, Osmolality, Percent Saturation, pH, Gamma Globulin, Potassium, IgG, SGPT, Sodium, Alkaline Phosphatase, TIBC, Total Bilirubin, Total Protein, Blood Urea Nitrogen, Calcium, Chloride, Creatinine, Gamma Globulin, Glucose, IgG, Insulin | |
| United States | |
| 500 mL | |
| Sequential-filtered, 100 nm | |
| Bovine Adenovirus, Bovine Parvovirus, Bovine RSV, BVDV, Cytopathogenic Agents, Hemadsorbing Agents, Rabies Virus, Reovirus | |
| ≤ 10 mg/dL |
| Sera should be stored at -10°C or lower. Once thawed, sera should be stored at 2° to 8°C. | |
| Bovine Growth Serum | |
| Sterile | |
| None | |
| ≤ 10 EU/mL | |
| No |