missing translation for 'onlineSavingsMsg'
Learn More

Cytiva HyClone™ Bovine Growth Serum (U.S.)

Catalog No. SH3054103HI
Change view
Click to view available options
Quantity:
500 mL
Processing:
Heat Inactivated
None
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Processing
SH3054103HI 500 mL Heat Inactivated
SH3054103 500 mL None
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. SH3054103HI Supplier Cytiva Supplier No. SH30541.03HI
Get 1 Consumable Product When You Buy 1 Obtenez 1 produit consommable à l’achat d’un produit
View all promotions

Support cell culture growth with U.S.-sourced HyClone™ Bovine Growth Serum, a superior performing fetal bovine serum (FBS) replacement, consisting of bovine calf serum supplemented with chemically-defined components.

  • Filtered through sequential 100nm (0.1μm) pore-size rated filters

Specifications

Additional Assays Albumin, Alkaline Phosphatase, Blood Urea Nitrogen, Calcium, Chloride, Creatinine, Gamma Globulin, Glucose, IgG, Insulin, Lactate Dehydrogenase, SGPT, SGOT, pH, Total Protein, Total Bilirubin, Sodium, Potassium, Phosphorus, Osmolality, Iron, TIBC, Percent Saturation, G
Content And Storage Sera should be stored at -10°C or lower. Once thawed, sera should be stored at 2° to 8°C.
Product Type Bovine Growth Serum
Quantity 500 mL
Sterility Sterile
Sterilization Method Sequential-filtered, 100 nm
Processing Heat Inactivated
Tested For Bluetongue, Bluetongue; Bovine; Adenovirus; Bovine Parvovirus; Bovine RSV; BVDV; Cytopathogenic Agents; Hemadsorbing Agents; Rabies Virus; Reovirus, Bovine Adenovirus, Bovine Parvovirus, Bovine RSV, BVDV, Cytopathogenic Agents, Hemadsorbing Agents, Rabies Virus, Reovirus
Endotoxin Concentration ≤ 10 EU/mL
Heat Inactivated Yes
Hemoglobin Concentration ≤ 10 mg/dL
Mycoplasma Detected Yes
Show More Show Less