missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva HyClone™ Bovine Growth Serum
A cost-effective alternative to Fetal Bovine Serum (FBS).
Supplier: Cytiva SH3054102HI
Specifications
| Insulin, Gamma Globulin, Alkaline Phosphatase, Lactate Dehydrogenase, SGPT, SGOT, pH, Total Protein, Albumin, Blood Urea Nitrogen, Creatinine, Total Bilirubin, Sodium, Potassium, Calcium, Chloride, Phosphorus, Osmolality, Iron, TIBC, Percent Saturation, Glucose, IgG | |
| Heat Inact.; 100 mL; Available as quote | |
| 100 mL | |
| Sequential-filtered, 100 nm | |
| Bovine Adenovirus, Bovine Parvovirus, Bovine RSV, BVDV, Cytopathogenic Agents, Hemadsorbing Agents, Rabies Virus, Reovirus | |
| Yes | |
| Yes |
| Sera should be stored at -10°C or lower. Once thawed, sera should be stored at 2° to 8°C. | |
| Bovine Growth Serum | |
| Sterile | |
| Heat Inactivated | |
| ≤ 10 EU/mL | |
| ≤ 10 mg/dL |