missing translation for 'onlineSavingsMsg'
Learn More

Cytiva HyClone™ Bovine Calf Serum (BCS), US Origin, Irradiated 25-40 kGy

Catalog No. SH3734IR254
Change view
Click to view available options
Quantity:
1000 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SH3734IR254 1000 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SH3734IR254 Supplier Cytiva Supplier No. SH30073.04IR2540
NLSU: Get 1 Consumable Product When You Buy 1 Obtenez 1 produit consommable à l’achat d’un produit
View all promotions

A cost-effective alternative to Fetal Bovine Serum (FBS).

  • Sourced in the USA with complete traceability back to original source
  • Cost-effective alternative to FBS
  • Age of animal at time of collection approximately 16-24 weeks
  • Low in antibodies and high in growth factors
  • Virus panel testing according to 9 CFR 113.53

Specifications

Content And Storage Sera should be stored at -10°C or lower. Once thawed, sera should be stored at 2°C to 8°C.
Country of Origin United States
Quantity 1000 mL
Sterilization Method Irradiated
Endotoxin Concentration ≤ 10 EU/mL
Hemoglobin Concentration < 30 mg/dL
Mycoplasma Detected Not Detected