Learn More
Cytiva HyClone™ Alpha Calf Fraction media supplement (U.S. origin)
Catalog No. SH3021203
Change view
Click to view available options
Description: Without Iron
Catalog No.
SH3021203 Description: Without Iron
1 options
Catalog No. SH3021203 Supplier Cytiva Supplier No. SH30212.03
NLSU: Get 1 Consumable Product When You Buy 1 Obtenez 1 produit consommable à l’achat d’un produit
View all promotions
Promote growth with this cost-effective media supplement, designed for antibody production in hybridoma cell cultures.
HyClone Alpha Calf Fraction is a cost-effective medium supplement that promotes growth and antibody production in hybridoma cell cultures. This product is produced from our bovine calf serum, using a non-ethanol proprietary process to reduce immunoglobulins and protein to levels very similar to those of fetal bovine serum (FBS). Alpha Calf Fraction typically contains less than 200 μg IgG /mL. Alpha Calf Fraction is filtered through three sequential 100 nm (0.1 µm) pore size-rated filters. Before dispensing, each lot of serum is pooled using true pool technology to ensure uniformity and consistency between bottles.
Specifications
Additional Assays Gamma Globulin,Alkaline Phosphatase,Lactate Dehydrogenase,SGPT,SGOT,pH,Albumin,Blood Urea Nitrogen,Creatinine,Total Bilirubin,Sodium,Potassium,Calcium,Chloride,Inorganic Phosphorus,Iron,TIBC,Percent Saturation,Glucose
Description Without Iron
Sterility Sterile
Sterilization Method Filtered through three sequential 100 nm (0.1 µm) pore size-rated filters