missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva HyClone™ AdvanceSTEM™ Mesenchymal Stem Cell Expansion Kit
Supplier: Cytiva SH3087902
Description
Support expansion and maintenance of undifferentiated human mesenchymal stem cells (hMSCs) with HyClone™ AdvanceSTEM™ Mesenchymal Stem Cell Expansion Kit.
Specifications
| Undifferentiated Human Mesenchymal Stem Cells | |
| Stem Cell Differentiation Media | |
| 1000Â mL | |
| Mesenchymal Stem Cell Basal Medium |
| Liquid | |
| Sterile-filtered | |
| Store at 2° to 8°C away from light |