missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva HyClone™ AdvanceSTEM™ ES qualified Hepes buffer
Specifications
Specifications
| Certifications/Compliance | cGMP and ISO 91:2 Guidelines |
| Content And Storage | 2° to 8°C |
| Description | AdvanceSTEM ES Qualified HEPES (1M) |
| Concentration | 1 M |
| Form | Liquid |
| Product Type | HEPES Buffer |
| Sterility | Sterile |
| Sterilization Method | 0.1 μm Sterile-filtered |
| Endotoxin Level | FIO |
| pH | 7.2 to 7.5 |
| Show More |