missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva HyClone™ ActiPro Powdered Cell Culture Media
Description
HyClone ActiPro cell culture medium is available in both liquid and powder forms for cell culture bioprocesses. This chemically defined medium is intended for use in combination with HyClone ActiSM medium and Cell Boost 7a and Cell Boost 7b supplements.
ActiPro media and supplements are formulated for industrial production of recombinant proteins, including those requiring complex post-translational modifications and correct protein folding. In addition, this engineered ActiPro system of media and supplements can significantly reduce production process time.
- Demonstrates high yields of recombinant protein
- Does not contain hypoxanthine and thymidine (HAT supplement)
- Chemically defined ADCF mAb media formulation
- Production medium for batch or fed-batch fermentation processes
Specifications
Specifications
| Form | Powder |
| Sterility | Non-sterile |
| With Additives | Poloxamer 188 |
| Without Additives | No Glutamine |
| Quantity | 1 L |
| Content And Storage | 2 to 8°C, away from light and moisture |