missing translation for 'onlineSavingsMsg'
Learn More

Cytiva™ HyClone™ ActiPro™ Cell Culture Media

Catalog No. SH3103901
Change view
Click to view available options
Quantity:
500 mL
1000 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SH3103901 500 mL
SH3103902 1000 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. SH3103901 Supplier Cytiva™ Supplier No. SH31039.01
NLSU: Get 1 Consumable Product When You Buy 1 Obtenez 1 produit consommable à l’achat d’un produit
View all promotions

Chemically defined ADCF cell culture medium providing high yields of recombinant proteins in processes using Chinese hamster ovary (CHO) cells.

HyClone™ ActiPro™ cell culture media has been formulated to provide high yields of recombinant proteins in bioprocesses using Chinese hamster ovary (CHO) cell lines. The cultured medium is chemically defined, animal-derived component-free (ADCF), and optimized for high-yield protein expression and production in batch or CHO fed-batch cell culture processes.

  • Demonstrates high yields of recombinant protein
  • Does not contain hypoxanthine and thymidine (HAT supplement)
  • Chemically defined ADCF mAb media formulation
  • Production medium for batch or fed-batch fermentation processes

HyClone™ ActiPro™ cell culture medium is available in both liquid and powder forms for cell culture bioprocesses. This chemically defined medium is intended for use in combination with HyClone™ ActiSM™ medium and Cell Boost™ 7a and Cell Boost™ 7b supplements. ActiPro™ media and supplements are formulated for industrial production of recombinant proteins, including those requiring complex post-translational modifications and correct protein folding. In addition, this engineered ActiPro™ system of media and supplements can significantly reduce production process time.

The ActiPro™ system is suitable for general cell culture biomanufacturing with CHO cell lines such as CHO-GS, CHO-K1, CHO-DG44, and CHO-S. ActiPro™ cell culture media and supplements were designed with adaptation and CHO protein production in mind. ActiPro™ media and supplements are balanced formulations of amino acids, vitamins, salts, trace elements, and other components, meeting the metabolic needs of CHO cells in production. The cell culture system does not contain peptides, hydrolysates, phenol red, 2-mercaptoethanol, or growth factors such as insulin, to ensure batch-to-batch consistency and increased cell culture bioprocess efficiency. For fast-growing CHO cells with higher carbon consumption, glucose supplementation is needed.

Specifications

Cell Line CHO
Cell Type CHO
Classification Protein Production
Form Liquid
Product Type Cell Culture Media
Sterility Sterile-filtered
Sterilization Method 0.2 μm Sterile-filtered
With Additives Poloxamer 188
Without Additives No Glutamine, No Thymidine, No Hypoxanthine
Product Line HyClone ActiPro
Quantity 500 mL
Culture Type Mammalian Cell Culture
Expression System Mammalian
Show More Show Less