missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva HyClone™ ActiCHO™ SM Powdered Media
Description
ActiCHO SM powdered cell culture medium is chemically defined, containing poloxamer 188, without insulin or L-glutamine.
- Hydrolysate-free and animal-derived component-free (ADCF). There are no proteins, peptides or growth factors in ActiCHO SM medium.
Specifications
Specifications
| Form | Powder |
| Sterility | Non-sterile |
| With Additives | Poloxamer 188 |
| Without Additives | No Glutamine, No Insulin |
| Quantity | 1 L |
| Content And Storage | 2 to 8°C, away from light and moisture |